Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ddost
Entrez Gene ID
13200
Gene Name
dolichyl-di-phosphooligosaccharide-protein glycotransferase
Gene Aliases
-
Chromosome Location
Chr.4: 138304738 - 138312611 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007838.2 NP_031864.2 7 - 8 947 78
GenBank mRNA AK077651.1 - 7 - 8 809 78
AK081542.1 - 7 - 8 839 78
AK084267.1 - 7 - 8 831 78
AK148095.1 - 7 - 8 828 78
AK150683.1 - 7 - 8 838 78
BC068132.1 - 7 - 8 947 78
D89063.1 - 7 - 8 949 78

View Full Product Details