Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gatad2a
Entrez Gene ID
234366
Gene Name
GATA zinc finger domain containing 2A
Gene Aliases
1110066C11Rik, BC031407, C80248
Chromosome Location
Chr.8: 69907069 - 69996384 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286450.1 NP_001273379.1 1 - 2 448 94
NM_145596.4 NP_663571.2 1 - 2 448 94
GenBank mRNA AK089074.1 - 1 - 2 430 94
AK160058.1 - 1 - 2 297 94
BB654622.1 - 1 - 2 443 94
CV562594.1 - 1 - 2 206 94

View Full Product Details