Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gatad2a
Entrez Gene ID
234366
Gene Name
GATA zinc finger domain containing 2A
Gene Aliases
1110066C11Rik, BC031407, C80248
Chromosome Location
Chr.8: 69907069 - 69996384 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113346.1 NP_001106817.1 2 - 3 396 76
NM_001286450.1 NP_001273379.1 2 - 3 544 76
NM_145596.4 NP_663571.2 2 - 3 544 76
GenBank mRNA AK083473.1 - 2 - 3 546 76
AK089074.1 - 2 - 3 526 76
AK160058.1 - 2 - 3 393 76
BB654622.1 - 2 - 3 539 76
BC038221.1 - 2 - 3 294 76
CV562594.1 - 2 - 3 302 76

View Full Product Details