The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001045484.1 | NP_001038949.1 | 5 - 6 | 610 | 101 |
NM_008578.2 | NP_032604.2 | 5 - 6 | 610 | 101 | |
GenBank mRNA | BC045147.1 | - | 5 - 6 | 610 | 101 |
CO811773.1 | - | - | 277 | 101 |