Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Crtc1
Entrez Gene ID
382056
Gene Name
CREB regulated transcription coactivator 1
Gene Aliases
AI413414, Mect1, R74955, TORC1, mKIAA0616
Chromosome Location
Chr.8: 70382352 - 70439643 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004062.2 NP_001004062.1 12 - 13 1515 57
GenBank mRNA AK129173.1 - 12 - 13 1512 57
BC056137.1 - 12 - 13 6103 57
BC068139.1 - 12 - 13 1521 57
BC080308.1 - 12 - 13 1515 57

View Full Product Details