The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040692.3 | NP_001035782.1 | 1 - 2 | 246 | 114 |
NM_001136087.2 | NP_001129559.1 | 1 - 2 | 246 | 114 | |
NM_001168643.1 | NP_001162114.1 | 1 - 2 | 246 | 114 | |
NM_001168644.1 | NP_001162115.1 | 1 - 2 | 246 | 114 | |
NM_001168645.1 | NP_001162116.1 | 1 - 2 | 246 | 114 | |
NM_001168646.1 | NP_001162117.1 | 1 - 2 | 246 | 114 | |
GenBank mRNA | AF075262.1 | - | 1 - 2 | 223 | 114 |
AF075263.1 | - | 1 - 2 | 223 | 114 | |
AF075264.1 | - | 1 - 2 | 223 | 114 | |
AF075265.1 | - | 1 - 2 | 223 | 114 | |
AF075266.1 | - | 1 - 2 | 223 | 114 | |
AF075267.1 | - | 1 - 2 | 223 | 114 | |
AK085258.1 | - | 1 - 2 | 243 | 114 | |
AK085473.1 | - | 1 - 2 | 244 | 114 | |
AK128907.1 | - | 1 - 2 | 243 | 114 | |
BC010748.1 | - | 1 - 2 | 222 | 114 | |
CA491609.1 | - | 1 - 2 | 283 | 114 |