The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016709.2 | NP_057918.2 | 6 - 7 | 618 | 69 |
GenBank mRNA | AK003219.1 | - | 6 - 7 | 598 | 69 |
BC026525.1 | - | 6 - 7 | 556 | 69 | |
BC049597.1 | - | 6 - 7 | 602 | 69 |