Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fastkd3
Entrez Gene ID
69577
Gene Name
FAST kinase domains 3
Gene Aliases
2310010B21Rik, AW048587
Chromosome Location
Chr.13: 68582163 - 68592338 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027123.4 NP_081399.3 4 - 5 1830 73
GenBank mRNA AK009264.1 - 4 - 5 589 73
AK031142.1 - 4 - 5 1817 73
AK033189.1 - 4 - 5 1831 73
AK148902.1 - 4 - 5 1821 73

View Full Product Details