Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rusc2
Entrez Gene ID
100213
Gene Name
RUN and SH3 domain containing 2
Gene Aliases
AI840675, mKIAA0375
Chromosome Location
Chr.4: 43381982 - 43427092 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037709.2 NP_001032798.1 8 - 9 3645 102
NM_199057.3 NP_951012.2 8 - 9 3606 102
GenBank mRNA AK122263.1 - 8 - 9 3525 102
AK132170.1 - 8 - 9 3535 102
AK166260.1 - 8 - 9 3546 102
BC024790.1 - 6 - 7 836 102
BC141186.1 - 7 - 8 3376 102

View Full Product Details