Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf38
Entrez Gene ID
73469
Gene Name
ring finger protein 38
Gene Aliases
1700065B19Rik, 2610202O07Rik, AA673263, Oip1
Chromosome Location
Chr.4: 44126212 - 44233643 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038993.3 NP_001034082.1 4 - 5 602 70
NM_175201.5 NP_780410.2 3 - 4 632 70
GenBank mRNA AK006888.1 - 3 - 4 594 70
AK147608.1 - 4 - 5 659 70
BC060730.1 - 3 - 4 609 70
BC062976.1 - 3 - 4 546 70

View Full Product Details