The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172122.1 | NP_001165593.1 | 10 - 11 | 1603 | 56 |
NM_001172123.1 | NP_001165594.1 | 10 - 11 | 1603 | 56 | |
NM_001172124.1 | NP_001165595.1 | 10 - 11 | 1603 | 56 | |
NM_001172126.1 | NP_001165597.1 | 10 - 11 | 1603 | 56 | |
GenBank mRNA | AK030609.1 | - | 10 - 11 | 1237 | 56 |
AK033005.1 | - | 10 - 11 | 1246 | 56 | |
AK041098.1 | - | 10 - 11 | 1605 | 56 | |
AK052196.1 | - | 10 - 11 | 1584 | 56 |