The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172121.1 | NP_001165592.1 | 13 - 14 | 1321 | 83 |
NM_001172122.1 | NP_001165593.1 | 12 - 13 | 1796 | 83 | |
NM_001172123.1 | NP_001165594.1 | 13 - 14 | 1844 | 83 | |
NM_178660.4 | NP_848775.2 | 12 - 13 | 1793 | 83 | |
GenBank mRNA | AK030478.1 | - | 12 - 13 | 1453 | 83 |
AK030609.1 | - | 12 - 13 | 1430 | 83 | |
AK032938.1 | - | 12 - 13 | 1790 | 83 | |
AK046197.1 | - | 13 - 14 | 1322 | 83 | |
AK052196.1 | - | 13 - 14 | 1825 | 83 | |
BC108402.1 | - | 12 - 13 | 1767 | 83 | |
BC117841.1 | - | 12 - 13 | 1233 | 83 |