Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vwa5b2
Entrez Gene ID
328643
Gene Name
von Willebrand factor A domain containing 5B2
Gene Aliases
BB182430, EG328644, Gm1738, Gm931
Chromosome Location
Chr.16: 20589319 - 20605378 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144953.1 NP_001138425.1 13 - 14 1914 60
NM_182636.4 NP_872574.2 13 - 14 1914 60
GenBank mRNA AK141799.1 - 13 - 14 1915 60

View Full Product Details