Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc14
Entrez Gene ID
239839
Gene Name
coiled-coil domain containing 14
Gene Aliases
G630039H03Rik
Chromosome Location
Chr.16: 34690567 - 34725204 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK029725.1 - 10 - 11 2908 178

View Full Product Details