Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vwa5b1
Entrez Gene ID
75718
Gene Name
von Willebrand factor A domain containing 5B1
Gene Aliases
4931403E03Rik
Chromosome Location
Chr.4: 138564229 - 138635937 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029401.1 NP_083677.1 6 - 7 924 55
GenBank mRNA AK016425.1 - 6 - 7 945 55
BC057376.1 - 5 - 6 796 55
BC132477.1 - 6 - 7 924 55
BC137851.1 - 6 - 7 924 55

View Full Product Details