The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163288.2 | NP_001156760.2 | 4 - 5 | 852 | 71 |
GenBank mRNA | BC157977.1 | - | 4 - 5 | 777 | 71 |
BC158124.1 | - | 4 - 5 | 829 | 71 |