Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Susd1
Entrez Gene ID
634731
Gene Name
sushi domain containing 1
Gene Aliases
A530080P10, Gm12528, OTTMUSG00000007332
Chromosome Location
Chr.4: 59314202 - 59439289 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163288.2 NP_001156760.2 12 - 13 2078 82
GenBank mRNA AK041075.1 - 3 - 4 356 82
BC125408.1 - 2 - 3 236 82
BC125410.1 - 2 - 3 236 82
BC157977.1 - 12 - 13 2003 82
BC158124.1 - 12 - 13 2055 82

View Full Product Details