The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023054.1 | NP_075541.1 | 1 - 1 | 1313 | 87 |
GenBank mRNA | AF155362.2 | - | 1 - 1 | 1308 | 87 |
AF271213.1 | - | 1 - 1 | 1313 | 87 | |
AK010316.1 | - | 1 - 1 | 1330 | 87 | |
AK145637.1 | - | 1 - 1 | 1326 | 87 | |
BC022994.1 | - | 1 - 1 | 1318 | 87 |