Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ctdspl
Entrez Gene ID
69274
Gene Name
CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase-like
Gene Aliases
2810418J22Rik, AI426263, HYA22, SCP3
Chromosome Location
Chr.9: 118926536 - 119044119 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133710.3 NP_598471.3 4 - 5 692 81
GenBank mRNA AA239542.1 - 4 - 5 383 81
AJ344340.1 - 4 - 5 774 81
AK013115.1 - 4 - 5 688 81
BC094289.1 - 4 - 5 377 81

View Full Product Details