Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl2
Entrez Gene ID
77113
Gene Name
kelch-like 2, Mayven
Gene Aliases
6030411N21Rik, ABP-KELCH, AU020744, Mav
Chromosome Location
Chr.8: 64739675 - 64793152 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178633.3 NP_848748.2 - 2015 97
GenBank mRNA AK032116.1 - - 1563 97
AK167903.1 - - 1806 97
BC024572.1 - - 1182 97
BC031142.1 - - 1886 97
BC031144.1 - - 1909 97

View Full Product Details