The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011817.2 | NP_035947.2 | 2 - 3 | 264 | 58 |
GenBank mRNA | AB021884.1 | - | 2 - 3 | 261 | 58 |
AF055638.1 | - | 2 - 3 | 252 | 58 | |
AK002237.1 | - | 2 - 3 | 262 | 58 | |
AK007410.1 | - | 2 - 3 | 267 | 58 | |
BC001989.1 | - | 2 - 3 | 245 | 58 |