The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026004.3 | NP_080280.3 | 1 - 2 | 265 | 110 |
GenBank mRNA | AK011525.1 | - | 1 - 2 | 216 | 110 |
AK011894.1 | - | 1 - 2 | 163 | 110 | |
BY080680.1 | - | 1 - 2 | 267 | 110 |