Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cylc2
Entrez Gene ID
74914
Gene Name
cylicin, basic protein of sperm head cytoskeleton 2
Gene Aliases
4930488P18Rik
Chromosome Location
Chr.4: 51216667 - 51229963 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162865.1 NP_001156337.1 2 - 3 86 120

View Full Product Details