The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081174.2 | NP_001074643.1 | 1 - 2 | 106 | 74 |
GenBank mRNA | BC147765.1 | - | - | 43 | 74 |
BC147767.1 | - | - | 43 | 74 |