The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012044.2 | NP_036174.1 | 1 - 2 | 131 | 92 |
GenBank mRNA | AF112984.1 | - | 1 - 2 | 131 | 92 |
AF166098.1 | - | 1 - 2 | 127 | 92 | |
AK148187.1 | - | - | 147 | 92 |