Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sytl4
Entrez Gene ID
27359
Gene Name
synaptotagmin-like 4
Gene Aliases
Gph, Slp4
Chromosome Location
Chr.X: 133936385 - 133981822 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290717.1 NP_001277646.1 3 - 4 538 110
NM_001290718.1 NP_001277647.1 3 - 4 511 110
NM_001290719.1 NP_001277648.1 3 - 4 538 110
NM_013757.2 NP_038785.1 3 - 4 538 110
GenBank mRNA AB025258.1 - 3 - 4 509 110
AB025259.1 - 3 - 4 509 110
AB120510.1 - 2 - 3 324 110
AK030401.1 - 3 - 4 385 110
AK161582.1 - 3 - 4 505 110
BC026819.1 - 3 - 4 538 110

View Full Product Details