Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Epha2
Entrez Gene ID
13836
Gene Name
Eph receptor A2
Gene Aliases
AW545284, Eck, Myk2, Sek-2, Sek2
Chromosome Location
Chr.4: 141301221 - 141329384 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010139.3 NP_034269.2 11 - 12 2189 92
GenBank mRNA AK137704.1 - 11 - 12 2171 92
AK144202.1 - 11 - 12 2173 92
BC140960.1 - 11 - 12 2145 92
U07634.1 - 11 - 12 2147 92
X76010.1 - 4 - 5 404 92
X78339.1 - 11 - 12 2151 92

View Full Product Details