Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Epha2
Entrez Gene ID
13836
Gene Name
Eph receptor A2
Gene Aliases
AW545284, Eck, Myk2, Sek-2, Sek2
Chromosome Location
Chr.4: 141301221 - 141329384 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010139.3 NP_034269.2 14 - 15 2608 59
GenBank mRNA AK137704.1 - 14 - 15 2590 59
AK144202.1 - 14 - 15 2592 59
BC140960.1 - 14 - 15 2564 59
X76010.1 - 7 - 8 823 59
X78339.1 - 14 - 15 2570 59

View Full Product Details