The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168472.1 | NP_001161944.1 | 1 - 2 | 258 | 151 |
GenBank mRNA | BY249698.1 | - | 1 - 2 | 258 | 151 |