Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem5
Entrez Gene ID
216395
Gene Name
transmembrane protein 5
Gene Aliases
6330415D21Rik
Chromosome Location
Chr.10: 122081120 - 122097366 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC083125.1 - 2 - 3 375 71

View Full Product Details