Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Taf10
Entrez Gene ID
24075
Gene Name
TAF10 RNA polymerase II, TATA box binding protein (TBP)-associated factor
Gene Aliases
30kDa, AU041226, TAFII30, Taf2h
Chromosome Location
Chr.7: 105742894 - 105744375 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020024.3 NP_064408.2 1 - 2 257 87
GenBank mRNA AK047115.1 - - 259 87
AK139383.1 - - 267 87
BC031418.1 - 1 - 2 257 87
BC080311.1 - 1 - 2 258 87

View Full Product Details