The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024919.1 | NP_001020090.1 | 3 - 4 | 679 | 94 |
GenBank mRNA | AK009485.1 | - | 3 - 4 | 661 | 94 |
AK208742.1 | - | 1 - 2 | 58 | 94 |