Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rhox2f
Entrez Gene ID
434764
Gene Name
reproductive homeobox 2F
Gene Aliases
EG434764, Rhox2.6, Rhox2e
Chromosome Location
Chr.X: 37571421 - 37576159 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085356.1 NP_001078825.1 1 - 2 91 120

View Full Product Details