Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp3a57
Entrez Gene ID
622127
Gene Name
cytochrome P450, family 3, subfamily a, polypeptide 57
Gene Aliases
EG622127
Chromosome Location
Chr.5: 145345145 - 145390926 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100180.1 NP_001093650.1 4 - 5 400 141

View Full Product Details