Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Krt83
Entrez Gene ID
406219
Gene Name
keratin 83
Gene Aliases
Krt2-25, Krt87
Chromosome Location
Chr.15: 101431490 - 101438804 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003668.2 NP_001003668.2 3 - 4 695 149
GenBank mRNA BC139321.1 - 3 - 4 695 149
BC139322.1 - 3 - 4 695 149

View Full Product Details