Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rps15a
Entrez Gene ID
267019
Gene Name
ribosomal protein S15A
Gene Aliases
A630031B11Rik
Chromosome Location
Chr.7: 118104374 - 118116147 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK010652.1 - 1 - 2 53 90

View Full Product Details