Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vmn2r80
Entrez Gene ID
624765
Gene Name
vomeronasal 2, receptor 80
Gene Aliases
EG624765
Chromosome Location
Chr.10: 79148816 - 79194933 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103368.1 NP_001096838.1 5 - 6 1670 140

View Full Product Details