Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vmn2r86
Entrez Gene ID
625109
Gene Name
vomeronasal 2, receptor 86
Gene Aliases
EG625109
Chromosome Location
Chr.10: 130446199 - 130455894 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103365.1 NP_001096835.1 4 - 5 1515 117

View Full Product Details