Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Msmp
Entrez Gene ID
100039672
Gene Name
microseminoprotein, prostate associated
Gene Aliases
OTTMUSG00000007017, Psmp
Chromosome Location
Chr.4: 43583216 - 43584494 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099314.1 NP_001092784.1 1 - 2 358 156

View Full Product Details