Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vmn2r99
Entrez Gene ID
665376
Gene Name
vomeronasal 2, receptor 99
Gene Aliases
EG665376
Chromosome Location
Chr.17: 19362135 - 19394590 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104551.2 NP_001098021.2 5 - 6 1638 143

View Full Product Details