Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vmn2r93
Entrez Gene ID
627132
Gene Name
vomeronasal 2, receptor 93
Gene Aliases
EG627132
Chromosome Location
Chr.17: 18298281 - 18326441 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104542.1 NP_001098012.1 5 - 6 1648 160

View Full Product Details