Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Btnl6
Entrez Gene ID
624681
Gene Name
butyrophilin-like 6
Gene Aliases
EG624681, Gm6519, NG13
Chromosome Location
Chr.17: 34507935 - 34517352 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030747.1 NP_109672.1 1 - 2 83 96

View Full Product Details