Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sprr1b
Entrez Gene ID
20754
Gene Name
small proline-rich protein 1B
Gene Aliases
-
Chromosome Location
Chr.3: 92436809 - 92438779 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009265.3 NP_033291.1 1 - 2 40 72
GenBank mRNA AK003398.1 - 1 - 2 41 72
BC119288.1 - 1 - 2 33 72
BC119290.1 - 1 - 2 33 72

View Full Product Details