Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Espn
Entrez Gene ID
56226
Gene Name
espin
Gene Aliases
je
Chromosome Location
Chr.4: 152120331 - 152152420 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019585.3 NP_062531.2 1 - 2 161 120
GenBank mRNA AK002284.1 - 1 - 2 122 120
BC044723.1 - 1 - 2 161 120
BC057888.1 - 1 - 2 107 120

View Full Product Details