The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144848.2 | NP_659097.2 | - | 1090 | 78 |
GenBank mRNA | AY312170.1 | - | - | 1090 | 78 |