The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207278.2 | NP_997161.1 | 1 - 1 | 3265 | 126 |
GenBank mRNA | AK081667.1 | - | 1 - 1 | 3267 | 126 |