Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Scaf8
Entrez Gene ID
106583
Gene Name
SR-related CTD-associated factor 8
Gene Aliases
A630086M08Rik, A930036P18Rik, AA408306, AI447644, AI448652, AU015035, Rbm16, mKIAA1116
Chromosome Location
Chr.17: 3114972 - 3198859 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK041620.1 - 1 - 1 2194 137
AK042381.1 - 4 - 4 1426 137

View Full Product Details