Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ctxn3
Entrez Gene ID
629147
Gene Name
cortexin 3
Gene Aliases
ENSMUSG00000069372
Chromosome Location
Chr.18: 57467656 - 57478134 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134697.1 NP_001128169.1 1 - 2 346 92
GenBank mRNA AK042789.1 - 1 - 2 306 92

View Full Product Details