Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Entpd4
- Entrez Gene ID
-
67464
- Gene Name
- ectonucleoside triphosphate diphosphohydrolase 4
- Gene Aliases
-
1200014F22Rik, 4930513O14Rik, A430102M06Rik, AI414893, Lysal1, mKIAA0392
- Chromosome Location
-
Chr.14: 69337149 - 69366742 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK015790.1 |
- |
- |
606 |
107 |
View Full Product Details