Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Btbd17
Entrez Gene ID
72014
Gene Name
BTB (POZ) domain containing 17
Gene Aliases
1500005I02Rik, Tango10a
Chromosome Location
Chr.11: 114790669 - 114802640 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC057599.1 - 1 - 2 150 85

View Full Product Details